Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Propionylpyridine 98.0+%, TCI America™
SDP

Catalog No. P20901G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P20901G 1 g
P20905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P20901G Supplier TCI America Supplier No. P20901G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1570-48-5
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.166
MDL Number MFCD00023589
InChI Key VDNKJMUNLKAGAM-UHFFFAOYSA-N
Synonym 3-propionylpyridine, 1-3-pyridyl propan-1-one, ethyl 3-pyridyl ketone, 1-pyridin-3-yl propan-1-one, 3-pyridyl ethyl ketone, 1-3-pyridinyl-1-propanone, 3-propionyl pyridine, 1-pyridin-3-yl-propan-1-one, ethyl pyridin-3-yl ketone, 1-propanone, 1-3-pyridinyl
PubChem CID 74074
IUPAC Name 1-pyridin-3-ylpropan-1-one
SMILES CCC(=O)C1=CN=CC=C1

Specifications

Color Yellow
Boiling Point 77°C
Quantity 1 g
Formula Weight 135.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Propionylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.