Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Propoxypropylamine 98.0+%, TCI America™
SDP

Catalog No. P10955ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P10955ML 5 mL
P109525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P10955ML Supplier TCI America Supplier No. P10955ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16728-59-9
Molecular Formula C6H15NO
Molecular Weight (g/mol) 117.19
MDL Number MFCD00129001
InChI Key UTOXFQVLOTVLSD-UHFFFAOYSA-N
Synonym 1-Amino-3-propoxypropane
PubChem CID 85579
IUPAC Name 3-propoxypropan-1-amine
SMILES CCCOCCCN

Specifications

Color Colorless
Boiling Point 152°C
Quantity 5 mL
UN Number 2733
Formula Weight 117.19
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Propoxypropylamine
EINECSNumber (2)-0385
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.