Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Pyrrolidin-1-ylmethyl)aniline, 97%, Thermo Scientific™

Catalog No. CC55714DA
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC55714DA 1 g
CC55714CB 250 mg
CC55714DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC55714DA Supplier Thermo Scientific Chemicals Supplier No. CC55714DA
Add to Cart

Chemical Identifiers

CAS 183365-31-3
Molecular Formula C11H16N2
Molecular Weight (g/mol) 176.263
InChI Key RZDBCURLSQNEAD-UHFFFAOYSA-N
Synonym 3-pyrrolidin-1-ylmethyl aniline, 3-pyrrolidin-1-ylmethyl-phenylamine, benzenamine, 3-1-pyrrolidinylmethyl, 3-1-pyrrolidinylmethyl aniline, 3-pyrrolidinylmethyl phenylamine, 3-pyrrolidinylmethylaniline, 3-pyrrolidin-1-ylmethyl-aniline, 3-1-pyrrolidinylmethyl benzenamine, 3-pyrrolidin-1-yl-methyl-aniline, 3-pyrrolidin-1-yl methyl aniline
PubChem CID 1096407
IUPAC Name 3-(pyrrolidin-1-ylmethyl)aniline
SMILES C1CCN(C1)CC2=CC(=CC=C2)N

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 176.26
Percent Purity 97%
Chemical Name or Material 3-(Pyrrolidin-1-ylmethyl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.