Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex 3 Ring Binder Holder/Pen Holder
SDP

Catalog No. 22000510
Change view
Click to view available options
Mounting Type:
Magnetic
VHB Tape
Wall
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting Type
22-000-510 Wall
22-000-511 Magnetic
22-000-512 VHB Tape
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-000-510 Supplier Poltex Supplier No. BINDRPENW
Add to Cart
Add to Cart

Super durable PETG construction.

  • Holds 3 ring binder and pens in side pocket.
  • Mounts with powerful magnets, VHB Tape or screws/anchors to wall.

Specifications

Product Type Binder Holder
Color Clear
Material PETG
Mounting Type Wall
Height (Metric) 23 cm
Length (Metric) 8 cm
Width (Metric) 31 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.