Learn More
Gilson Company Inc 3"SIEVE BR/SS FULL #60
Supplier:  Gilson Company Inc V3CF #60
Encompass_Preferred
3" Sieve, Brass/Stainless, Full Height, No. 603in (76mm) diameter ASTM E11 Test Sieve has stainless steel woven-wire cloth with No. 60 (250µm) openings and full height brass frame. The overall height is 1-3/4in and the stacked height is 1-1/8in.
Catalog No. NC2147394
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.