Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(tert-Butyl)-1-methyl-1H-pyrazole-carboxylic acid, 95%

Catalog No. AC452620050
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC452620050 5 g
AC452620500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC452620050 Supplier Thermo Scientific Chemicals Supplier No. 452620050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 175277-11-9 | C9H14N2O2 | 182.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 175277-11-9
Molecular Formula C9H14N2O2
Molecular Weight (g/mol) 182.22
InChI Key SFSXXMXHJOSBAZ-UHFFFAOYSA-N
Synonym 3-tert-butyl-1-methyl-1h-pyrazole-5-carboxylic acid, 3-tert-butyl-1-methylpyrazole-5-carboxylic acid, 5-tert-butyl-2-methyl-2h-pyrazole-3-carboxylic acid, 1h-pyrazole-5-carboxylic acid, 3-1,1-dimethylethyl-1-methyl, 3-t-butyl-1-methyl-1h-pyrazole-5-carboxylic acid, maybridge1_008671, acmc-1bzn5, butylmethylpyrazolecarboxylicacid
PubChem CID 720529
IUPAC Name 5-tert-butyl-2-methylpyrazole-3-carboxylic acid
SMILES CC(C)(C)C1=NN(C(=C1)C(=O)O)C

Specifications

Melting Point 151.0°C to 154.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 182.22
Percent Purity 95%
Chemical Name or Material 3-(tert-Butyl)-1-methyl-1H-pyrazole-carboxylic acid
Hazard Category Warning
Hazard Statement GHS H Statement Causes skin irritation. May cause respiratory irritation. Causes serious eye irritation.
Precautionary Statement GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.