Learn More
3-tert-Butyltoluene, 99%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Melting Point | -41°C |
| Boiling Point | 189°C |
| Odor | Aromatic |
| Quantity | 1 g |
| UN Number | UN2667 |
| Formula Weight | 148.25 |
| Percent Purity | 99% |
| Chemical Name or Material | 3-tert-Butyltoluene |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement H331-H227-H302-H319-H335 Toxic if inhaled. Combustible liquid. Harmful if swallowed. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement P210-P261-P280-P305+P351+P338-P405-P501a Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BUTYLTOLUENES |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.