Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(3-Thien-3-ylphenyl)methanol, ≥97%, Thermo Scientific™

Catalog No. CC48409DA
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC48409DA 1 g
CC48409CB 250 mg
CC48409DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC48409DA Supplier Thermo Scientific Chemicals Supplier No. CC48409DA
Add to Cart

Chemical Identifiers

CAS 89929-82-8
Molecular Formula C11H10OS
Molecular Weight (g/mol) 190.26
MDL Number MFCD06203083
InChI Key VSWBZMMHNMDWJL-UHFFFAOYSA-N
Synonym 3-thien-3-ylphenyl methanol, 3-thiophen-3-yl phenyl methanol, 3-3-thienyl phenyl methanol, 3-thiophen-3-ylphenyl methanol, acmc-20ls08, 3-3-thienyl phenylmethanol, 3-3-thiophenyl phenyl methanol, 3-3-thienyl phenyl methan-1-ol
PubChem CID 7164662
IUPAC Name (3-thiophen-3-ylphenyl)methanol
SMILES C1=CC(=CC(=C1)CO)C2=CSC=C2

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 190.27
Percent Purity 95%
Chemical Name or Material (3-Thien-3-ylphenyl)methanol
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.