Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Thiophenecarboxaldehyde, 98%

Catalog No. AC187980250
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC187980250 25 mL Glass bottle
AC187980050 5 mL Glass bottle
AC187981000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC187980250 Supplier Thermo Scientific Chemicals Supplier No. 187980250
Add to Cart

CAS: 498-62-4 | C5H4OS | 112.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 498-62-4
MDL Number MFCD00005466
InChI Key RBIGKSZIQCTIJF-UHFFFAOYSA-N
Synonym 3-thiophenecarboxaldehyde, thiophene-3-carboxaldehyde, 3-formylthiophene, 3-thenaldehyde, 3-formyl-thiophene, 3-thiophenealdehyde, thiophene-3-aldehyde, 3-thiophene carboxaldehyde, 3-thiophenecarbaldehyde, 3-thiophenaldehyde
PubChem CID 68135
ChEBI CHEBI:87611
IUPAC Name thiophene-3-carbaldehyde
SMILES C1=CSC=C1C=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -30.0°C
Color Yellow to Brown
Density 1.2800g/mL
Boiling Point 194.0°C to 196.0°C
Flash Point 64°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C5H4OS
Refractive Index 1.5820 to 1.5860
Quantity 25 mL
Specific Gravity 1.28
Molecular Weight (g/mol) 112.146
Formula Weight 112.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Thiophenecarboxaldehyde, 98%
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
May cause an allergic skin reaction.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 207-865-5
TSCA TSCA
Recommended Storage Product may darken on standing

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.