Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Trifluoromethoxy)benzaldehyde 97.0+%, TCI America™
SDP

Catalog No. T182425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T182425G 25 g
T18245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T182425G Supplier TCI America Supplier No. T182425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 52771-21-8
Molecular Formula C8H5F3O2
Molecular Weight (g/mol) 190.12
MDL Number MFCD00042406
InChI Key FQEVHRCPXFKJHF-UHFFFAOYSA-N
Synonym 3-trifluoromethoxy benzaldehyde, m-trifluoromethoxy benzaldehyde, m-trifluoromethoxybenzaldehyde, pubchem1470, acmc-209l1m, 3trifluoromethoxybenzaldehyde, 3-trifluormethoxy-benzaldehyde, ksc274m0j, 3-trifluoromethoxy-benzaldehyde, timtec-bb sbb006601
PubChem CID 605139
IUPAC Name 3-(trifluoromethoxy)benzaldehyde
SMILES FC(F)(F)OC1=CC=CC(C=O)=C1

Specifications

Color Yellow
Quantity 25 g
Formula Weight 190.12
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(Trifluoromethoxy)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.