Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Trifluoromethoxy)cinnamic Acid 98.0+%, TCI America™
SDP

Catalog No. T178025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T178025G 25 g
T17805G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T178025G Supplier TCI America Supplier No. T178025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 168833-80-5
Molecular Formula C10H7F3O3
Molecular Weight (g/mol) 232.158
MDL Number MFCD00066337
InChI Key CLKZZEYGXRWYNI-SNAWJCMRSA-N
Synonym 3-trifluoromethoxy cinnamic acid, 3-3-trifluoromethoxy phenyl acrylic acid, trans-3-trifluoromethoxy cinnamic acid, 2e-3-3-trifluoromethoxy phenyl prop-2-enoic acid, rarechem bk hw 0216, 3-3-trifluoromethoxy-phenyl-acrylic acid, e-3-3-trifluoromethoxy phenyl acrylic acid, e-3-3-trifluoromethoxy phenyl prop-2-enoic acid, 3-3-trifluoromethoxy phenyl prop-2-enoic acid, pubchem4353
PubChem CID 735840
IUPAC Name (E)-3-[3-(trifluoromethoxy)phenyl]prop-2-enoic acid
SMILES C1=CC(=CC(=C1)OC(F)(F)F)C=CC(=O)O

Specifications

Melting Point 95°C
Color White-Yellow
Quantity 25 g
Formula Weight 232.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(Trifluoromethoxy)cinnamic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.