Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3'-(Trifluoromethyl)acetophenone 97.0+%, TCI America™
SDP

Catalog No. T154625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T154625G 25 g
T15465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T154625G Supplier TCI America Supplier No. T154625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 349-76-8
Molecular Formula C9H7F3O
Molecular Weight (g/mol) 188.149
MDL Number MFCD00000391
InChI Key ABXGMGUHGLQMAW-UHFFFAOYSA-N
Synonym 3'-trifluoromethyl acetophenone, 3-trifluoromethyl acetophenone, 3-trifluoromethylacetophenone, ethanone, 1-3-trifluoromethyl phenyl, 1-3-trifluoromethyl phenyl ethanone, m-trifluoromethylacetophenone, 3'-trifluoromethylacetophenone, 1-3-trifluoromethyl phenyl ethan-1-one, 1-3-trifluoromethyl-phenyl-ethanone
PubChem CID 67682
IUPAC Name 1-[3-(trifluoromethyl)phenyl]ethanone
SMILES CC(=O)C1=CC(=CC=C1)C(F)(F)F

Specifications

Boiling Point 199°C
Quantity 25 g
Formula Weight 188.15
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3′-(Trifluoromethyl)acetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.