Learn More
3-(Trifluoromethyl)aniline, 99%
Description
Used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Used as pharmaceutical intermediate.
Solubility
Soluble in water. [5 g/L (20°C)]
Notes
Store in cool dry place. Keep away from oxidizing agents.
Specifications
Specifications
| Melting Point | 5°C to 6°C |
| Density | 1.295 |
| Boiling Point | 187°C to 189°C |
| Flash Point | 85°C (185°F) |
| Odor | Recognizable |
| Refractive Index | 1.48 |
| Quantity | 2500 g |
| UN Number | UN2948 |
| Beilstein | 387672 |
| Solubility Information | Soluble in water. [5g/L (20°C)] |
| Show More |
Safety and Handling
| Hazard Category | H227-H302+H312-H315-H318-H330-H335-H373 |
| Hazard Statement | GHS H Statement H330-H373-H227-H302-H312-H319 Fatal if inhaled. May cause damage to organs through prolonged or repeated exposure. Combustible liquid. Harmful if swallowed. Harmful in contact with skin. Causes serious eye irritation. |
| Precautionary Statement | P210-P235-P260-P264b-P270-P271-P280-P284-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P314-P330-P332+P313-P362-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: 3-TRIFLUOROMETHYLANILINE |
| EINECSNumber | 202-643-4 |
| RTECSNumber | XU9180000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.