Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Trifluoromethyl)benzyl Bromide 98.0+%, TCI America™
SDP

Catalog No. T178225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T178225G 25 g
T17825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T178225G Supplier TCI America Supplier No. T178225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 402-23-3
Molecular Formula C8H6BrF3
Molecular Weight (g/mol) 239.04
MDL Number MFCD00000395
InChI Key MYYYZNVAUZVXBO-UHFFFAOYSA-N
Synonym 3-trifluoromethyl benzyl bromide, 1-bromomethyl-3-trifluoromethyl benzene, m-trifluoromethylbenzyl bromide, 3-trifluoromethylbenzyl bromide, 3-bromomethyl benzotrifluoride, 3-trifluoromethylbenzylbromide, 3-trifluoromethyl benzylbromide, 1-bromomethyl-3-trifluoromethyl-benzene, alpha'-bromo-alpha,alpha,alpha-trifluoro-m-xylene
PubChem CID 123061
IUPAC Name 1-(bromomethyl)-3-(trifluoromethyl)benzene
SMILES FC(F)(F)C1=CC=CC(CBr)=C1

Specifications

Color Yellow
Boiling Point 79°C
Quantity 25 g
UN Number 3265
Formula Weight 239.04
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(Trifluoromethyl)benzyl Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.