Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Trifluoromethyl)benzyl chloride, 98%

Catalog No. AAA1625314
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1625314 25 g
AAA1625306 5 g
AAA1625322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1625314 Supplier Thermo Scientific Chemicals Supplier No. A1625314
Add to Cart

CAS: 705-29-3 | C8H6ClF3 | 194.581 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 705-29-3
Molecular Formula C8H6ClF3
Molecular Weight (g/mol) 194.581
MDL Number MFCD00000908
InChI Key XGASTRVQNVVYIZ-UHFFFAOYSA-N
Synonym 3-trifluoromethyl benzyl chloride, 1-chloromethyl-3-trifluoromethyl benzene, 3-trifluoromethylbenzyl chloride, 3-chloromethyl benzotrifluoride, m-trifluoromethylbenzyl chloride, 3-chloromethyl-benzotrifluoride, benzene, 1-chloromethyl-3-trifluoromethyl, alpha'-chloro-alpha,alpha,alpha-trifluoro-m-xylene, 3-chloromethylbenzotrifluoride, 3-trifluoromethylbenzylchloride
PubChem CID 69716
IUPAC Name 1-(chloromethyl)-3-(trifluoromethyl)benzene
SMILES C1=CC(=CC(=C1)C(F)(F)F)CCl

Specifications

Density 1.254
Boiling Point 68°C to 70°C (12 mmHg)
Flash Point 74°C (165°F)
Odor Pungent
Refractive Index 1.4645
Quantity 25 g
UN Number UN3265
Beilstein 743242
Formula Weight 194.59
Percent Purity 98%
Chemical Name or Material 3-(Trifluoromethyl)benzyl chloride
Show More Show Less
Hazard Category H226-H314
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 211-884-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.