Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Trifluoromethyl)phenylacetic Acid 98.0+%, TCI America™
SDP

Catalog No. T166225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T166225G 25 g
T16625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T166225G Supplier TCI America Supplier No. T166225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 351-35-9
Molecular Formula C9H7F3O2
Molecular Weight (g/mol) 204.148
MDL Number MFCD00004339
InChI Key BLXXCCIBGGBDHI-UHFFFAOYSA-N
Synonym 3-trifluoromethyl phenylacetic acid, m-trifluoromethyl phenylacetic acid, 2-3-trifluoromethyl phenyl acetic acid, benzeneacetic acid, 3-trifluoromethyl, 3-trifluoromethylphenylacetic acid, 3-trifluoromethyl phenyl acetic acid, a,a,a-trifluoro-m-tolyl acetic acid, alpha,alpha,alpha-trifluoro-m-tolyl acetic acid, 3-carboxymethyl benzotrifluoride
PubChem CID 67695
IUPAC Name 2-[3-(trifluoromethyl)phenyl]acetic acid
SMILES C1=CC(=CC(=C1)C(F)(F)F)CC(=O)O

Specifications

Melting Point 77°C
Color White-Yellow
Quantity 25 g
Formula Weight 204.15
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(Trifluoromethyl)phenylacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.