Learn More
Sigma Organic Chemistry 3- trifluoromethyl pyrr 2 | 25mg | MFCD03452798
Supplier: Sigma Organic Chemistry CDS01039125MG
3- trifluoromethyl pyrr 2, 25mg
About this Item:
MDL #: MFCD03452798
Molecular Weight: 175.58
UNSPSC Code: 12352200
Chemical Formula: C5H9ClF3N
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.