Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Trimethylsilyl)-1-propanesulfonic acid, sodium salt hydrate, 99+%, Thermo Scientific™

Catalog No. AC176650050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC176650050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC176650050 Supplier Thermo Scientific Chemicals Supplier No. 176650050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2039-96-5
Molecular Formula C6H15NaO3SSi
Molecular Weight (g/mol) 218.32
MDL Number MFCD00007537
InChI Key HWEXKRHYVOGVDA-UHFFFAOYSA-M
Synonym sodium 3-trimethylsilyl propane-1-sulfonate, 3-trimethylsilyl-1-propanesulfonic acid sodium salt, dss sodium salt, sodium 3-trimethylsilyl-1-propanesulfonate, 1-propanesulfonic acid, 3-trimethylsilyl-, sodium salt, 2,2-dimethyl-2-silapentane-5-sulfonate sodium salt, sodium 3-sulfopropyltrimethylsilane, 1-propanesulfonic acid, 3-trimethylsilyl-, sodium salt 1:1, sodium 2,2-dimethyl-2-silapentane-5-sulfonate, 3-trimethylsilyl propanesulfonic acid
PubChem CID 5167273
IUPAC Name sodium 3-(trimethylsilyl)propane-1-sulfonate
SMILES [Na+].C[Si](C)(C)CCCS([O-])(=O)=O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Color White to Beige
Elemental Analysis 99.0% min.
Infrared Spectrum Authentic
Assay Percent Range 99+%
Linear Formula (CH3)3Si(CH2)3SO3Na·xH2O
Quantity 5 g
Formula Weight 218.31
Physical Form Crystalline Powder
Chemical Name or Material 3-(Trimethylsilyl)-1-propanesulfonic acid, sodium salt hydrate, 99+%
Show More Show Less
Hazard Category HYGROSCOPIC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.