Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Trimethylsilyl)-2-naphthyl Trifluoromethanesulfonate 95.0+%, TCI America™
SDP

Catalog No. T24665G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24665G 5 g
T24661G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T24665G Supplier TCI America Supplier No. T24665G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 780820-43-1
Molecular Formula C14H15F3O3SSi
Molecular Weight (g/mol) 348.411
InChI Key OSZYFPWWHQEAGW-UHFFFAOYSA-N
Synonym 3-(Trimethylsilyl)-2-naphthyl Triflate, Trifluoromethanesulfonic Acid 3-(Trimethylsilyl)-2-naphthyl Ester
PubChem CID 44629848
IUPAC Name (3-trimethylsilylnaphthalen-2-yl) trifluoromethanesulfonate
SMILES C[Si](C)(C)C1=CC2=CC=CC=C2C=C1OS(=O)(=O)C(F)(F)F

Specifications

Color Yellow
Quantity 5 g
UN Number 3265
Formula Weight 348.41
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(Trimethylsilyl)-2-naphthyl Trifluoromethanesulfonate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.