Learn More
United Scientific Supplies 3-Way Glass Adapters
SDPSCI_ED
Catalog No. S43783 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Includes:
105° Sidearm
75° Sidearm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
S43783 105° Sidearm
S43782 75° Sidearm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S43783 Supplier United Scientific Supplies Supplier No. ADB10524
Add to Cart
Add to Cart
Makes connecting glassware safe and easy
  • Adapters have 24/40 joints
Specifications
Description Outer Joints on the Sidearm and Top; Inner Joint on the Bottom
Includes 105° Sidearm
Material Borosilicate Glass
Product Type 3-Way Adapter
Standard Taper Joint No. 24/40