Learn More
Justrite™ 30 Gallon ChemCor™ Corrosives/Acids Safety Cabinet With Bi-Fold Self-Closing Doors
Shop All Justrite Safety Group Products
Description
The ChemCor™ thermoplastic coating on all interior surfaces provides superior protection against corrosion from drips and off-gassing from harsh chemicals. Seam-free surface eliminates the need for rivets or fasteners which risk corrosion from seep-through chemicals. Non-porous surface resists staining and cleans up easily.
- Lid opens to no more than 60 degrees and stays closed when not in use, isolating contents from fire sources and limiting oxygen to virtually eliminate the risk of spontaneous combustion
- Round construction and elevated base allow circulation of air around can to disperse heat and Reduce moisture build-up and rusting
- Durable steel construction with a powder-coat finish provides chemical resistance
- Approved by FM, UL, and TUV
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Color | Blue |
| Capacity (English) | 30 gal. |
| Capacity (Metric) | 114 L |
| Material | ChemCor |