Learn More
Novus Biologicals 300 UL VIAL INSERT 100/PKG
Supplier: Novus Biologicals 045135
300 uL vial insert 100/pkg. Suitable for use with both iCE3 configurations and iCE280-alcott 719 configuration.