Learn More
Bullard™ 3000 Series Patriot Safety Helmet
SDP
Catalog No. 19084462 Shop All Bullard Company Products
Change view
Click to view available options
Suspension Type:
6-Point Ratchet or Pin-Lock
Pinlock suspension
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Suspension Type
19-084462 Pinlock suspension
19-084463 6-Point Ratchet or Pin-Lock
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-084462 Supplier Bullard™ Supplier No. TT1
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
The 3000 Series Patriot Safety Helmet is ideal for almost any industrial application, and its American Flag design will let you proudly demonstrate your patriotism. Permanent full graphic coverage will not fade, peel, or crack. Choose the six-point ratchet or pinlock suspension to provide a comfortable and secure fit. Accessory slots accept faceshields, hearing protection, welding helmets, or communication devices. Replaceable Sportek™ wicking browpad comes standard. Also available: replacement ratchet and pinlock suspensions, and replacement browpads.
Compliance
Meets ANSI Z89.1-1997 Type I, Class E, G and C requirements.
Specifications
Certifications/Compliance Meets ANSI Z89.1-1997 Type I; Class E, G and C.
Suspension Type Pinlock suspension
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.