Learn More
McMaster-Carr 304 SS CAP 6IN TUBE HP QC SAN
Supplier:  McMaster-Carr 4322K226
Encompass
The McMaster-Carr 304 Stainless Steel Strut Channel Cover is made from 304 stainless steel for corrosion resistance. It has a satin finish and features a snap-on design that installs by snapping over strut channels up to 1-5/8" wide to provide an aesthetic, finished look. The covers are available in 10 ft lengths with widths of 1", 1-1/4", 1-1/2", and 1-5/8". They can be cut to length with a hacksaw. These strut channel covers provide a clean, professional appearance.
Catalog No. NC2457733
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.