Learn More
Justrite™ 31 Gallon ChemCor™ Under Fume Hood Corrosives/Acids Safety Cabinet With Manual-Closing Doors
GSA_VAGREENER_CHOICE
Catalog No. 19822378 Shop All Justrite Safety Group Products
Change view
Click to view available options
Color:
Blue
Light Neutral
Silver
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-822-378 Light Neutral
19-822-377 Blue
19-822-380 Silver
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-822-378 Supplier Justrite™ Supplier No. 8849072
Add to Cart
Add to Cart
Request a Quote
Increase workplace safety and improve cleaniliness with the proper safety and storage equipment. The Justrite™ 31 Gallon ChemCor™ Under Fume Hood Corrosives/Acids Safety Cabinet With Manual-Closing Doors provides superior corrosion protection as well as fire protection in a single cabinet.
  • Expand storage, add new convenience to existing cabinets, or customize your new cabinet with additional shelves to meet specific workflow needs
  • Galvanized steel shelves hold up to 250 lbs and adjust on 3 in. increments for versatile storage
  • Patented SpillSlope galvanized steel shelves direct spills to back and bottom of a leakproof sump
  • Polyethylene accessories with a slightly curved edge provide additional protection against leaks and spills
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Load Bearing Capacity 31 gal.
Door Style Self-close
Color Light Neutral