Learn More
Justrite™ 31 Gallon Under Fume Hood Solvent/Flammable Liquid Safety Cabinet With Manual-Closing Doors
GSA_VAGREENER_CHOICE
Catalog No. 19822346 Shop All Justrite Safety Group Products
Change view
Click to view available options
Color:
Light Neutral
Silver
Yellow
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-822-346 Light Neutral
19-822-347 Silver
19-822-348 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-822-346 Supplier Justrite™ Supplier No. 884807
Only null left
Add to Cart
Add to Cart
Request a Quote
Increase workplace safety and improve cleaniliness with the proper safety and storage equipment. The Justrite™ 31 Gallon Under Fume Hood Solvent/Flammable Liquid Safety Cabinet With Manual-Closing Doors is fully compliant and designed for safe storage of solvents and flammables in laboratories.
  • Low-Profile EcoPolyBlend Accumulation Centers only measure 5-1/2 in. high for easy handling of drums
  • Made of recycled polyethylene, these accumulation centers are nearly impervious to chemicals, including acids and corrosives
  • Seamless construction eliminates leaks, making these ideal for drum filling and dispensing operations
  • Self-locating grates won't ride-up, and are removable for easy cleaning
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Load Bearing Capacity 31 gal.
Door Style Manual-close
Color Light Neutral