Learn More
Justrite™ 31 Gallon Under Fume Hood Solvent/Flammable Liquid Safety Cabinet With Self-Closing Doors
GSA_VAGREENER_CHOICE
Catalog No. 19822349 Shop All Justrite Safety Group Products
Change view
Click to view available options
Color:
Light Neutral
Silver
Yellow
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-822-349 Light Neutral
19-822-350 Silver
19-822-351 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-822-349 Supplier Justrite™ Supplier No. 884827
Add to Cart
Add to Cart
Request a Quote
Increase workplace safety and improve cleaniliness with the proper safety and storage equipment. The Justrite™ 31 Gallon Under Fume Hood Solvent/Flammable Liquid Safety Cabinet With Self-Closing Doors is fully compliant and designed for safe storage of solvents and flammables in laboratories.
  • Use these funnels with any closed-head 30- and 55-gallon drums
  • Low-profile, wide diameter opening makes it ideal for quick, easy pouring
  • Scalloped, sloped design minimizes splashes and prevents containers from resting in their own liquid
  • Large, flat well at the throat of the funnel allows for passive draining of paint tins, filters, buckets and other containers
  • Funnel for flammable liquids includes a drum fill vent with brass flame arrester, threaded bung cap with tight seal, and pressure relief valve for high temperatures
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Door Style Self-close
Load Bearing Capacity 31 gal.
Color Light Neutral