Learn More
Sigma Aldrich (Methylcyclopentadienyl)Manganese(I) Tricarbonyl
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
Specifications
Specifications
| CAS | 12108-13-3 |
| Melting Point | -1°C |
| Density | 1.38 g/mL at 25°C |
| Boiling Point | 232°C to 233°C |
| Molecular Formula | C9H7MnO3 |
| Linear Formula | C5H4CH3Mn(CO)3 |
| Refractive Index | n20/D 1.584 |
| MDL Number | MFCD00001426 |
| Quantity | 1 g |
| Synonym | (Methylcyclopentadienyl)tricarbonylmanganese; MCMT; Tricarbonyl(2-methylcyclopentadienyl)manganese; Ethyl MMT; MMT; NSC 22316 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.