Learn More
HOSOKAWA MICRON POWDER SYSTEMS HOSOKAWA 325 Mesh Test Sieve, ASTM E11 Certified, Durable Stainless Steel Frame, 200 mm Diameter
Supplier:  HOSOKAWA MICRON POWDER SYSTEMS M390003808CERT
Encompass
The HOSOKAWA M390-003808-CERT is a high-performance 325 mesh test sieve for precise particle size analysis, compatible with Mikro Air Jet Sieve MAJSx2. It features a 200 mm stainless steel frame and 45 µm pore size mesh. Certified to ASTM E11 standards, it ensures accuracy above 99%. Each sieve includes a Certificate of Conformance, a Certified Histogram, and is individually packed, providing reliable results for scientific applications.
Catalog No. NC1004196
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.