Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Fluorophenylmagnesium Bromide Solution

Catalog No. 328820100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
4 x 25 mL
800 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-882-0100ML 100 mL
32-882-025ML 4 x 25 mL
32-882-0800ML 800 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 32-882-0100ML Supplier Sigma Aldrich Supplier No. 328820100ML
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4-Fluorophenylmagnesium bromide solution (Grignard reagent) can be used as a reagent in the synthesis of:Useful key intermediate in the synthesis of paroxetine.Arylsulfides via cross-coupling reaction with thiols.4-Arylmethyl-1-phenylpyrazole and 4-aryloxy-1-phenylpyrazole derivatives as androgen receptor antagonists.Grignard intermediate for preparing aprepitant.

Specifications

CAS 352-13-6
Density 1.021 g/mL at 25°C
Molecular Formula C6H4BrFMg
Linear Formula FC6H4MgBr
MDL Number MFCD00009667
Quantity 100 mL
Molecular Weight (g/mol) 199.3 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.