Learn More
Arbor Assays 3,3-Cyclic GAMP ELISA Kit, (1 Plate)
SDP
Supplier:  Arbor Assays K073H1
Encompass_Preferred
The DetectX 3,3-cGAMP ELISA Kits is the World’s First ELISA Kit to measure 3,3-cGAMP in cell lysates, culture medium, and microbiome model systems and now has 10X increased sensitivity. This kit accurately quantifies 3’,3-cGAMP in bacterial or eukaryotic cell lysates. This innovative and valuable tool will facilitate further understanding of the role cyclic dinucleotides play in health and disease.
Catalog No. 50-227-8516
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.