Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pelican Products 3315C Safety Approved Flashlight

Catalog No. 19819200 Shop All Pelican Products Inc. Products
Encompass_Preferred
Change view
Click to view available options
Color:
Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-819-200 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options

Safety Certifications: CIDI, CIIDI, CIIIDI, IECEx ia

Catalog No. 19-819-200 Supplier Pelican Products Supplier No. 0331500103245

Safety Certifications: CIDI, CIIDI, CIIIDI, IECEx ia

The lightweight, compact design houses 3AA batteries that power a super bright LED light.

  • 3 AA Batteries (Included)
  • Built Pelican tough
  • Safety-approved for the most volatile work environments.
  • Class I, Class II, Class III – Division 1 Safety Certified
  • Safety Certified Class I, Division 1 / IECEx ia
  • Lanyard (included)
  • IP67 waterproof and dustproof
  • Removable clip included

Specifications

Light Source Single LED
Voltage 4.5 V
Weight (English) 3.7 oz. (without batteries)
Weight (Metric) 105 g (without batteries)
Battery Run Time 19 Hours
Battery Type 3 AA
Beam Distance 144 m
Certifications/Compliance CIDI, CIIDI, CIIIDI, IECEx ia
Color Yellow
Controller Type Push button on/off
Description Safety Approved/Certified
Diameter (English) Body 0.83 in.
Diameter (English) Head 1.57 in.
Diameter (Metric) Body 21 mm
Diameter (Metric) Head 40 mm
Dimensions (L x W) 6.14 x 1.57
Dimensions (L x W x H) 156 x 40 x 40 mm
For Use With (Application) Hazardous Work Enviroments
Includes 3 AA Batteries, Lanyard, Removable Clip
Length (English) 6.14 in.
Length (Metric) 15.6 cm
Lens Material Polycarbonate
Lumen 160 lm
Material ABS
Warranty Limited Lifetime Warranty
Product Type Flashlight
Show More Show Less

Compact LED Power, High Performance LED, 3 AA Batteries included. Built Pelican Tough, Lanyard, IP67 Waterproof, Removeable Clip, Accessories: Helment Mount, Nylon Holster

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.