Learn More
Sigma Aldrich Fine Chemicals Biosciences 3355-Tetramethylbenzidine
Supplier:  Sigma Aldrich Fine Chemicals Biosciences T5569100ML
Encompass
3 3AE 5 5AE-Tetramethylbenzidine (TMB) is an electrochemical substrate for low level detection of horse radish peroxidase (HRP). It is a highly sensitive and dual function substrate since it produces both soluble and insoluble reaction products. It has rapid reaction rate and it is suitable for on-line kinetic analysis. It does not possess carcinogenic properties.
Catalog No. 50-290-0896
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.