Learn More
Sigma Aldrich Fine Chemicals Biosciences 3355-Tetramethylbenzidine
Supplier:  Sigma Aldrich Fine Chemicals Biosciences T0565100ML
Encompass
3 3I 5 5I -Tetramethylbenzidine (TMB) is a chromogenic substrate for horseradish peroxidase (HRP) conjugates. It develops a permanent insoluble dark blue reaction product and is useful in colorimetric quantification.
Catalog No. 50-290-1879
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.