Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyldiphenylphosphine

Catalog No. 3369042G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-690-42G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 33-690-42G Supplier Sigma Aldrich Supplier No. 3369042G
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Catalyst for:Three component coupling reactions of arylaldehydes with Me vinyl ketone and phthalimideRegio- and stereoselective hydroalkynylation of methylenecyclopropanesSynthesis of oxazolidines, thiazolidines, pyrrolidines, and indolesDimer to monomer conversionTandem Morita-Baylis-Hillman/Michael addition reactionsPlatinum-catalyzed cyclizationRegioselective and stereoselective [3+2] cycloadditionPlatinum-catalyzed intermolecular hydroaminationHydroformylation reactions

Specifications

CAS 522-66-7
Melting Point 168°C to 176°C
Molecular Formula C20H26N2O2
Refractive Index n20/D 1.614
Linear Formula C20H26N2O2
MDL Number MFCD00151107
Quantity 5 g
Synonym Diphenylethylphosphine; NSC 151254
Molecular Weight (g/mol) 326.43 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.