Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Zirconium(IV) ethoxide

Catalog No. 33912125G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-912-125G 25 g
33-912-15G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 33-912-125G Supplier Sigma Aldrich Supplier No. 33912125G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Convenient precursor for the synthesis of olefin-functionalized zirconium-containing monomers, which have potential for the generation of hybrid zirconium-containing polymers.

Specifications

CAS 18267-08-8
Boiling Point 235°C (5 mmHg)
Molecular Formula C8H20O4Zr
Linear Formula Zr(OC2H5)4
Quantity 25 g
MDL Number MFCD00015654
Synonym Tetraethoxyzirconium; Zirconium ethoxide (Zr(OEt)4); Zirconium tetraethoxide
Molecular Weight (g/mol) 271.47
Percent Purity 97%
Biological Activity Respiratory system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.