Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sulfuric Acid

Catalog No. 339741500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-974-1500ML 500 mL
33-974-125L 2.5 L
33-974-1100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 33-974-1500ML Supplier Sigma Aldrich Supplier No. 339741500ML
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sulphuric acid may be prepared by catalytic oxidation of sulphur dioxide. It is a very strong electrolyte and has high affinity to water.Sulfuric acid can be used as a dehydrating agent, catalyst, and active reactant in the chemical industry. It is also used to prepare silica-sulfuric acid composite, which is used as a catalyst in organic synthesis.

Specifications

CAS 7664-93-9
Boiling Point ∼290°C
Density 1.840 g/mL at 25°C
Quantity 500 mL
Molecular Formula H2O4S
Linear Formula H2SO4
MDL Number MFCD00064589
Molecular Weight (g/mol) 98.08 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.