Learn More
Medchemexpress LLC HY-W017434 500mg Medchemexpress, 3,4-Dimethylbenzoic acid CAS:619-04-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW017434500MG
Encompass_Preferred
Medchemexpress, HY-W017434 500mg 3,4-Dimethylbenzoic acid CAS:619-04-5 3,4-Dimethylbenzoic acid acts as a product of dimethylbenzoate metabolism by Rhodococcus rhodochrous N75. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3410
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.