Learn More
Organomation 34 Position N-EVAP Nitrogen Evaporator
SDP
Supplier:  Organomation 11634
Encompass_Preferred
Our 34-position nitrogen evaporator (cat# 11634) offers a large number of sample positions, without compromising any of the classic N-EVAP features. This time-tested, durable system provides one of the least expensive prices per position of all our N-EVAPs. The evaporative system, along with the standard flow control and pressure reducing regulator, enables the operator to achieve the desired level of concentration with efficient nitrogen usage. Our unique dual band spring hoist assembly ensures easy and effective operator use.
Catalog No. NC9706907
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.