Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,4-Dibromobutyryl chloride

Catalog No. 3407210ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-072-10ML 10 mL
34-072-50ML 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 34-072-10ML Supplier Sigma Aldrich Supplier No. 3407210ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2,4-dibromobutyryl chloride was used in the preparation of:
  • Methyl 2,4-dibromobutanoate
  • Series of 3-(3, 5-di-tert-butyl-4-hydroxybenzylidene)pyrrolidin-2-ones, anti-inflammatory/analgesic agents
  • α-bromolactam

Specifications

CAS 82820-87-9
Density 2.00 g/mL (at 20°C)
Molecular Formula C4H5Br2ClO
Refractive Index n20/D 1.535
Linear Formula BrCH2CH2CHBrCOCl
MDL Number MFCD01862994
Quantity 10 mL
Molecular Weight (g/mol) 264.34
Percent Purity ∼90% (GC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.