Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cerium(Iv) Sulfate

Catalog No. 359009100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-900-9100G 100 g
35-900-925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 35-900-9100G Supplier Sigma Aldrich Supplier No. 359009100G
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant for:Preparation of spherical-like nanoparticles CeO2 nanoparticles for NO reductions reactionsOxidizing agentmay contain excess sulfate

Specifications

CAS 13590-82-4
Density 3.01 g/mL at 25°C
Molecular Formula CeO8S2
Linear Formula Ce(SO4)2
MDL Number MFCD00148852
Quantity 100 g
Synonym Ceric sulfate; Cerium disulfate
Molecular Weight (g/mol) 332.24 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.