Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium Dichloroisocyanurate Dihydrate

Catalog No. 3591550G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-915-50G 50 g
35-915-250G 250 g
35-915-1KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Warning
Catalog No. 35-915-50G Supplier Sigma Aldrich Supplier No. 3591550G
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium dichloroisocyanurate dihydrate is a solid chlorine bleaching agent generally used as an alternative to sodium hypochlorite for water treatment.Sodium dichloroisocyanurate dihydrate can be used as a chlorine source to convert alkoxyl-s-triazines into N-chlorinated alkoxy-s-triazines.

Specifications

CAS 51580-86-0
Molecular Formula C3Cl2N3NaO3 -+ 2H2O
Linear Formula C3Cl2N3NaO3 -+ 2H2O
MDL Number MFCD00149404
Quantity 50 g
Synonym NaDCC; Dichloroisocyanuric acid sodium salt dihydrate
Molecular Weight (g/mol) 255.98 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.