Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dibenzyl N,N-diethylphosphoramidite

Catalog No. 3628835G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-288-35G 5 g
36-288-31G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 36-288-35G Supplier Sigma Aldrich Supplier No. 3628835G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Dibenzyl n,n-diethylphosphoramidite is repoted to be suitable reagent for the efficient 1h-tetrazole-catalyzed αphosphite-triesterα phosphorylation of the protected serine derivatives.dibenzyl n,n-diethylphosphoramidite may be used:
  • As phosphitylating reagent in the synthesis of glycosyl phosphites and phosphates
  • For phosphorylating the alcohols
  • In the preparation of 6-dibenzylphosphate-1,3,5-tri-o-pmb-myo-inositol

Specifications

CAS 67746-43-4
Density 1.06 g/mL (at 25°C (literature))
Molecular Formula C18H24NO2P
Refractive Index n20/D 1.545 (literature)
Linear Formula (C2H5)2 NP(OCH2C6H5)2
MDL Number MFCD00191987
Quantity 5 g
Synonym TTC
Molecular Weight (g/mol) 317.36
Percent Purity 85%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.