Learn More
Sigma Organic Chemistry 3 4-Dihydroxybenzaldehy | 100G | 139-85-5 | MFCD00003370
Supplier: Sigma Organic Chemistry 37520100G
3 4-Dihydroxybenzaldehy , 100G
About this item:
CAS #: 139-85-5
MDL #: MFCD00003370
Purity: 0.97
Chemical Formula: C7H6O3
Molecular Weight: 138.12
UNSPSC Code: 12352100
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.