Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,3-Dichloro-2-(chloromethyl)-2-methylpropane

Catalog No. 38077625ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-077-625ML 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 38-077-625ML Supplier Sigma Aldrich Supplier No. 38077625ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,3-Dichloro-2-(chloromethyl)-2-methylpropane

Specifications

CAS 1067-09-0
Melting Point 18°C (lit.)
Density 1.271 g/mL (at 25°C (literature))
Boiling Point 89°C to 90°C (22 mmHg)
Molecular Formula C5H9Cl3
Refractive Index n20/D 1.482 (literature)
Linear Formula CH3C(CH2Cl)3
MDL Number MFCD00013686
Quantity 100 mg
Synonym 1,1,1-Tris(chloromethyl)ethane
Molecular Weight (g/mol) 175.48
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.