Learn More
Thermo Scientific™ Nunc™ 384-Well Polypropylene Sample Processing & Storage Microplates
GREENER_CHOICE
Catalog No. 12565384
Change view
Click to view available options
Color:
Black
Blue
Natural
White
Sterility:
Non-sterile
Sterile
Well Shape:
Square, Conical Bottom (V-well)
Square, Round Bottom (U-well)
Square-Rounded
Fit for Purpose:
Certified Quality
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Sterility Well Shape Fit for Purpose
12-565-384 Natural Non-sterile Square, Round Bottom (U-well) Certified Quality
12-565-385 Natural Sterile Square, Round Bottom (U-well) Certified Quality
12-565-386 White Non-sterile Square, Conical Bottom (V-well) Certified Quality
12-565-387 Black Non-sterile Square, Round Bottom (U-well) Certified Quality
12-565-390 Blue Non-sterile Square, Round Bottom (U-well) Certified Quality
14-377-859 Natural Non-sterile Square-Rounded Certified Quality
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 12-565-384 Supplier Thermo Scientific™ Supplier No. 264573
Only null left
Add to Cart
Add to Cart
Save on costs when storing compounds for small volume applications. Plates feature lower binding capacity so proteins or DNA will not bind, allowing for complete sample recovery.
Minimize costs when storing compounds for small volume applications. Thermo Scientific™ 384-Well Conical Bottom Polypropylene Plates feature a lower binding capacity so proteins or DNA will not bind, allowing for complete sample recovery. Withstands temperatures from -80° to +121°C. Offered in a variety of colors for quick identification of your storage plates.
  • Conical Wells are for optimal sample recovery
  • Square rounded wells minimize wicking (capillary action)
  • The variety of colors is for quick identification of storage plates
  • Chemical resistant to DMSO
TRUSTED_SUSTAINABILITY
Specifications
Barcode No Barcode
Certifications/Compliance Certified RNase and DNase Free. ANSI Compliant.
Format Standard
Lid Without Lid
Description 384-well Polypropylene Sample Processing & Storage Plates
Array 16 x 24
Autoclavable Autoclavable
Packaging 120 Cases
Binding Type Low binding PP
Product Line Nunc
Color Natural
Product Type 384-well plate
For Use With (Equipment) ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers
Material Polypropylene
Volume (Metric) Well 120 μL
No. of Wells 384
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Square, Round Bottom (U-well)
Dnase Free Yes
For Use With (Application) Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation
No. per Case 120
No. per Pack 20
Quantity 120/Cs.
Regulatory Status For Research Use Only. Not for use in diagnostic procedures.
RNase Free Yes
Fit for Purpose Certified Quality
Plate Size ANSI/ SLAS Standard