Learn More
Applied Biosystems™ Plate Septa, 384 well
Description
Includes
Each package contains 20 septa, sufficient for 7,680 samples
These are septa for use with the 3730, 3730xl, 3130xl, 3130, 3100 and 3100-Avant™ instruments.
Use with:
384-Well Plate Base and 384-Well Plate Retainer
For Research Use Only. Not for use in diagnostics procedures.
Specifications
Specifications
| Quantity | 20 each |
| Format | 384-well Plate |
| For Use With (Equipment) | 3100-Avant Genetic Analyzer |
| Product Type | Septa Plate |
| Shipping Condition | Room Temperature |
| Content And Storage | Each package contains 20 septa, sufficient for 7,680 samples. |
Frequently Asked Questions (FAQs)
We officially recommend that the plate septa be used one time only. However, we are aware that some of our customers will rinse the septa with water and re-use them. In those instances, the septa can become sticky and gummy over time and eventually start sticking to the array. In the best case scenario, the array will lift the plate off of the autosampler just enough to break the connection with the plate sensor and abort the run. The Event Log might show that the run aborted due to the user aborting it, so the run will appear to have completed or stopped with no errors and no data. The worst case scenario is that the plate remains on the array as the autosampler starts to move, bending the electrodes and breaking the array. We recommend replacing the plate septa regularly (replacing after each run is recommended). Do not use bleach, detergents, or solvents to clean the septa, it will cause them to dry out faster and also risks contaminating the system.
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.