Learn More
Greiner Bio-One 384 Well Polypropylene Flat Bottom Non-Sterile Microplates
Catalog No. 07000882 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Black
Natural
White
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07-000-882 White
07-000-049 Natural
07-000-883 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 07-000-882 Supplier Greiner Bio-One Supplier No. 781207
Add to Cart
Add to Cart
Resistant to many standard laboratory chemicals such as DMSO. Greiner Bio-One 384 Well Polypropylene Flat Bottom Non-Sterile Microplates are compatible with standard equipment and automated systems.
  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic
  • Manufactured under DIN ISO 9001 guidelinesNumber of wells are quadrupledWell-to-well spacing is 4.5mm
  • Rounded square well profile
  • Non-ste
Service Plan
No
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid Without Lid
Array 16 x 24
Binding Type Non Binding
Color White
Material Polypropylene
No. of Wells 384
Volume (Metric) Working 15 to 145 μL
Sterility Non-sterile
Total Volume (Metric) Well 152 μL
Well Shape Rounded Square
No. per Case 100
No. per Pack 10
Well Color White