Learn More
Greiner Bio-One 384-well Polypropylene Flat Bottom Sterile Microplate
Catalog No. 07000884 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Clear
Catalog No. Color
07-000-884 Clear
1 options
Catalog No. 07-000-884 Supplier Greiner Bio-One Supplier No. 781261
Add to Cart
Add to Cart
Resistant to many standard laboratory chemicals such as DMSO. Greiner Bio-One 384 Well Polypropylene Flat Bottom Sterile Microplates are compatible with standard equipment and automated systems.
  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic
TRUSTED_SUSTAINABILITY
Specifications
Array 16 x 24
Autoclavable Not Autoclavable
Color Clear
Material Polypropylene
No. of Wells 384
Volume (Metric) Working 15 to 145 μL
Sterility Sterile
Total Volume (Metric) Well 152 μL
Well Shape Rounded Square
Dnase Free Yes
Max. RCF 4800 x g (swinging bucket rotor)
No. per Case 100
RNase Free Yes
Temperature Range (Metric) -196°C to +121°C