Learn More
Greiner Bio-One 384-well Small Volume™ HiBase Polystyrene Microplates
Shop All Greiner Bio-One ProductsDescription
- Non-Pyrogenic
- Material: Polystyrene
- Free of detectable DNase, RNase, human DNA
- Traced way back to production through defined LOT number system
- Working volume: 4μL - 25μL
Certifications
DIN ISO 9001
Specifications
Specifications
| Format | Standard |
| Array | 16 x 24 |
| Color | Black |
| Material | Polystyrene |
| No. of Wells | 384 |
| Sterility | Sterile |
| No. per Case | 40 |
| No. per Pack | 10 |